missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-111984
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
89-84-9
C8H8O3
152.15
MFCD00002279
SULYEHHGGXARJS-UHFFFAOYSA-N
2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
6990
CHEBI:18414
CC(=O)C1=CC=C(O)C=C1O
This item is not returnable. View return policy
89-84-9
C8H8O3
152.15
MFCD00002279
SULYEHHGGXARJS-UHFFFAOYSA-N
2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
6990
CHEBI:18414
CC(=O)C1=CC=C(O)C=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 143.0°C to 147.0°C
Color Brown to White
Density 1.1800g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H3COCH3
Quantity 5 g
Beilstein 08, 266
Merck Index 15, 8260
Specific Gravity 1.18
Solubility Information Solubility in water: 1g/L (20 c). Other solubilities: soluble in glacial acetic acid,soluble in most common organic solvents except,benzene,chloroform and ether
IUPAC Name 1-(2,4-dihydroxyphenyl)ethanone
Formula Weight 152.15
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2', 4'-Dihydroxyacetophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2',4'-Dihydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'