missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorotoluene, 98%, Thermo Scientific™

Catalog Number p-110842
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
452-76-6
C7H6F2
128.12
MFCD00000332
MPXDAIBTYWGBSL-UHFFFAOYSA-N
67983
2,4-difluoro-1-methylbenzene
CC1=C(C=C(C=C1)F)F
This item is not returnable. View return policy
452-76-6
C7H6F2
128.12
MFCD00000332
MPXDAIBTYWGBSL-UHFFFAOYSA-N
67983
2,4-difluoro-1-methylbenzene
CC1=C(C=C(C=C1)F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1500g/mL
Boiling Point 114.0°C to 116.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3F2
Refractive Index 1.4475 to 1.4495
Quantity 5 g
Specific Gravity 1.15
Synonym 2,4-difluorotoluene, difluorotoluene, benzene, 2,4-difluoro-1-methyl, 2,4-difluorotolune, 2, 4-difluorotoluene, 2,4-difluoro methyl benzene, 2,4-difluoro-1-methyl-benzene, 2,4-difluor-1-methylbenzol, pubchem1612, pubchem1613
Formula Weight 128.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorotoluene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Difluorotoluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'