missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetone, 98%, Thermo Scientific Chemicals

Catalog Number p-3523291
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
274682-91-6
C9H8F2O
170.16
MFCD02258866
XVHHDTNIOCSTLH-UHFFFAOYSA-N
1-2,4-difluorophenyl propan-2-one, 2,4-difluorophenylacetone, 2-propanone, 1-2,4-difluorophenyl, 2,4-difluorophenyl propanone, 1-2,4-difluorophenyl-2-propanone, 1-2,4-difluoro-phenyl-propane-2-one
2734881
1-(2,4-difluorophenyl)propan-2-one
CC(=O)CC1=C(C=C(C=C1)F)F
This item is not returnable. View return policy
274682-91-6
C9H8F2O
170.16
MFCD02258866
XVHHDTNIOCSTLH-UHFFFAOYSA-N
1-2,4-difluorophenyl propan-2-one, 2,4-difluorophenylacetone, 2-propanone, 1-2,4-difluorophenyl, 2,4-difluorophenyl propanone, 1-2,4-difluorophenyl-2-propanone, 1-2,4-difluoro-phenyl-propane-2-one
2734881
1-(2,4-difluorophenyl)propan-2-one
CC(=O)CC1=C(C=C(C=C1)F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (F)2C6H3CH2COCH3
Refractive Index 1.4770 to 1.4790
Quantity 1 g
Formula Weight 170.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorophenylacetone
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 2,4-Difluorophenylacetone
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'