missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-2689778
21.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
72235-52-0
C7H7F2N
143.14
MFCD00010142
QDZZDVQGBKTLHV-UHFFFAOYSA-N
2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma
2733244
(2,4-difluorophenyl)methanamine
C1=CC(=C(C=C1F)F)CN
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
72235-52-0
C7H7F2N
143.14
MFCD00010142
QDZZDVQGBKTLHV-UHFFFAOYSA-N
2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma
2733244
(2,4-difluorophenyl)methanamine
C1=CC(=C(C=C1F)F)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 82°C to 84°C (15.0mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CH2NH2
Refractive Index 1.4900 to 1.493
Quantity 1 g
Specific Gravity 1.2
Formula Weight 143.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorobenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'