missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzenesulfonyl chloride, 98%, ACROS Organics™

Brand:  Acros Organics 357490250

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 13918-92-8 Weight : 0.07500kg Transport : UN number : 3265 Chem class : 1a Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10453405

  • £135.30 / 25g


Estimated Shipment
19-07-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

2, 4-Difluorobenzenesulfonyl chloride
97.5
≥97.5% (Akt. Cl)
C6H3ClF2O2S
212.6
13918-92-8
100.0
98%
(F)C6H3SOCl
MFCD00042481

Specifications

2, 4-Difluorobenzenesulfonyl chloride
Clear colorless to light yellow
13918-92-8
100.0
Glass bottle
Liquid
98%
(F)C6H3SOCl
MFCD00042481
1.581g/mL
25g
97.5
Authentic
1.5165 to 1.5185 (20°C, 589nm)
≥97.5% (Akt. Cl)
C6H3ClF2O2S
212.6

Safety and Handling

Product Identifier
  • 2,4-Difluorobenzenesulfonyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

Documentation

Technical Documents