missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzeneboronic acid 1,3-propanediol ester, 98%, Thermo Scientific™

Catalog Number p-7053823
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1073372-07-2
C9H9BF2O2
197.976
MFCD11504975
AHKNGRYTXDKKDQ-UHFFFAOYSA-N
2-2,4-difluorophenyl-1,3,2-dioxaborinane, 2,4-difluorophenylboronic acid, propanediol cyclic ester, 2,4-difluorophenylboronic acid propanediol cyclic ester, 2,4-difluorophenylboronic acid,propanediol cyclic ester, 2,4-difluorobenzeneboronic acid 1,3-propanediol ester
46739109
2-(2,4-difluorophenyl)-1,3,2-dioxaborinane
B1(OCCCO1)C2=C(C=C(C=C2)F)F
This item is not returnable. View return policy
1073372-07-2
C9H9BF2O2
197.976
MFCD11504975
AHKNGRYTXDKKDQ-UHFFFAOYSA-N
2-2,4-difluorophenyl-1,3,2-dioxaborinane, 2,4-difluorophenylboronic acid, propanediol cyclic ester, 2,4-difluorophenylboronic acid propanediol cyclic ester, 2,4-difluorophenylboronic acid,propanediol cyclic ester, 2,4-difluorobenzeneboronic acid 1,3-propanediol ester
46739109
2-(2,4-difluorophenyl)-1,3,2-dioxaborinane
B1(OCCCO1)C2=C(C=C(C=C2)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 197.98
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,4-Difluorobenzeneboronic acid 1,3-propanediol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'