missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoroaniline, 99%, Thermo Scientific Chemicals

Catalog Number 10645532
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
367-25-9
C6H5F2N
129.11
MFCD00007648
CEPCPXLLFXPZGW-UHFFFAOYSA-N
2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine
9709
2,4-difluoroaniline
NC1=CC=C(F)C=C1F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
367-25-9
C6H5F2N
129.11
MFCD00007648
CEPCPXLLFXPZGW-UHFFFAOYSA-N
2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine
9709
2,4-difluoroaniline
NC1=CC=C(F)C=C1F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -7.5°C
Color Brown
Density 1.2680g/mL
Boiling Point 170°C (753.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3NH2
Refractive Index 1.5025 to 1.5075
Quantity 25 mL
Beilstein 12, I, 297
Merck Index 15, 3166
Specific Gravity 1.268
Solubility Information Solubility in water: immiscible
Formula Weight 129.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluoroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Difluoroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'