missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoroacetophenone, 99%, Thermo Scientific Chemicals

Catalog Number p-3778270
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
364-83-0
C8H6F2O
156.12
MFCD00151261
QEWHNJPLPZOEKU-UHFFFAOYSA-N
2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213
67770
1-(2,4-difluorophenyl)ethanone
CC(=O)C1=C(C=C(C=C1)F)F
This item is not returnable. View return policy
364-83-0
C8H6F2O
156.12
MFCD00151261
QEWHNJPLPZOEKU-UHFFFAOYSA-N
2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213
67770
1-(2,4-difluorophenyl)ethanone
CC(=O)C1=C(C=C(C=C1)F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 80.0°C to 81.0°C (25.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3COCH3
Refractive Index 1.4870 to 1.4890
Quantity 10 g
Specific Gravity 1.23
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble/miscible in polar solvents,insoluble in non-polar solvents
Formula Weight 156.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2', 4'-Difluoroacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2',4'-Difluoroacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'