missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone, 97%, Thermo Scientific Chemicals

Catalog Number p-7054032
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
86404-63-9
C10H7F2N3O
223.183
MFCD02093825
XCHRPVARHBCFMJ-UHFFFAOYSA-N
1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethanone, 2,4-difluoro-alpha-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1,2,4-triazol-1-yl ethanone, 2',4'-difluoro-2-1h-1,2,4-triazol-1-yl acetophenone, unii-hxi8r9r915, 2-1h-1,2,4-triazol-1-yl-2',4'-difluoroacetophenone, ethanone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl, 2',4'-difluoro-2-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethan-1-one, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl-ethanone
588080
1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
C1=CC(=C(C=C1F)F)C(=O)CN2C=NC=N2
This item is not returnable. View return policy
86404-63-9
C10H7F2N3O
223.183
MFCD02093825
XCHRPVARHBCFMJ-UHFFFAOYSA-N
1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethanone, 2,4-difluoro-alpha-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1,2,4-triazol-1-yl ethanone, 2',4'-difluoro-2-1h-1,2,4-triazol-1-yl acetophenone, unii-hxi8r9r915, 2-1h-1,2,4-triazol-1-yl-2',4'-difluoroacetophenone, ethanone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl, 2',4'-difluoro-2-1h-1,2,4-triazolyl acetophenone, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl ethan-1-one, 1-2,4-difluorophenyl-2-1h-1,2,4-triazol-1-yl-ethanone
588080
1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
C1=CC(=C(C=C1F)F)C(=O)CN2C=NC=N2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 103°C to 107°C
Color Brown to White
Quantity 5 g
Formula Weight 223.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'