missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorotoluene, 99%, Thermo Scientific Chemicals

Catalog Number p-106243
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
95-73-8
C7H6Cl2
161.03
MFCD00000583
FUNUTBJJKQIVSY-UHFFFAOYSA-N
2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
7254
CHEBI:81651
2,4-dichloro-1-methylbenzene
CC1=C(C=C(C=C1)Cl)Cl
This item is not returnable. View return policy
95-73-8
C7H6Cl2
161.03
MFCD00000583
FUNUTBJJKQIVSY-UHFFFAOYSA-N
2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
7254
CHEBI:81651
2,4-dichloro-1-methylbenzene
CC1=C(C=C(C=C1)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -14.0°C
Color Colorless
Density 1.2400g/mL
Boiling Point 200.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3Cl2
Refractive Index 1.5450 to 1.5470
Quantity 250 mL
Beilstein 05, 295
Specific Gravity 1.24
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 161.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4-Dichlorotoluene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichlorotoluene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'