missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyridine, 98%, Thermo Scientific Chemicals

Catalog Number 11956331
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
26452-80-2
C5H3Cl2N
147.99
MFCD00955618
TYPVHTOETJVYIV-UHFFFAOYSA-N
pyridine, 2,4-dichloro, 2,4-dichloro pyridine, 2,4-dichoropyridine, 2, 4-dichloropyridine, 2,4-dichloro-pyridine, 2,4-dichlorpyridin, 2,4-dichlorpyridine, 2,4-dichloropyrdine, pubchem3531, acmc-1cnvt
33515
2,4-dichloropyridine
C1=CN=C(C=C1Cl)Cl
This item is not returnable. View return policy
26452-80-2
C5H3Cl2N
147.99
MFCD00955618
TYPVHTOETJVYIV-UHFFFAOYSA-N
pyridine, 2,4-dichloro, 2,4-dichloro pyridine, 2,4-dichoropyridine, 2, 4-dichloropyridine, 2,4-dichloro-pyridine, 2,4-dichlorpyridin, 2,4-dichlorpyridine, 2,4-dichloropyrdine, pubchem3531, acmc-1cnvt
33515
2,4-dichloropyridine
C1=CN=C(C=C1Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -1.0°C
Density 1.3700g/mL
Boiling Point 189.0°C to 190.0°C
Flash Point 89°C
Assay Percent Range 97.5% min. (GC)
Refractive Index 1.5500 to 1.5540
Quantity 100 g
Specific Gravity 1.37
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2, 4-Dichloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'