missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2',4'-Dichloropropiophenone, 97%

Brand:  Alfa Aesar™ A11252.14

Code : NEW

Additional Details : CAS Number : 37885-41-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11403604

  • £42.40 / 25g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Description

Description

Applications
2′,4′-Dichloropropiophenone is used in the synthesis of (S)-3-chloro-1-(4-chlorophenyl)-1-propanol.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

2',4'-Dichloropropiophenone
C9H8Cl2O
MFCD00027396
1-(2,4-Dichlorophenyl)-1-propanone
37885-41-9
203.07
2248270

Specifications

2',4'-Dichloropropiophenone
37885-41-9
C9H8Cl2O
MFCD00027396
2248270
1-(2,4-Dichlorophenyl)-1-propanone
25g
97%
203.07
121° to 123°C (7mmHg)
>110°C (230°F)
Videos
SDS