missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-2689772
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
68716-47-2
C6H5BCl2O2
190.81
MFCD00013930
QNEGDGPAXKYZHZ-UHFFFAOYSA-N
2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726, 2,4-dichloro benzene boronic acid, boronic acid, 2,4-dichlorophenyl, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronicacid
2734659
(2,4-dichlorophenyl)boronic acid
OB(O)C1=CC=C(Cl)C=C1Cl
This item is not returnable. View return policy
68716-47-2
C6H5BCl2O2
190.81
MFCD00013930
QNEGDGPAXKYZHZ-UHFFFAOYSA-N
2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726, 2,4-dichloro benzene boronic acid, boronic acid, 2,4-dichlorophenyl, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronicacid
2734659
(2,4-dichlorophenyl)boronic acid
OB(O)C1=CC=C(Cl)C=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 240.0°C to 245.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 190.82
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 4-Dichlorophenylboronic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichlorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'