missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenol, 99%, Thermo Scientific Chemicals

Catalog Number 10786341
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
120-83-2
C6H4Cl2O
163.00
MFCD00002169
HFZWRUODUSTPEG-UHFFFAOYSA-N
4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
8449
CHEBI:16738
2,4-dichlorophenol
OC1=CC=C(Cl)C=C1Cl
120-83-2
C6H4Cl2O
163.00
MFCD00002169
HFZWRUODUSTPEG-UHFFFAOYSA-N
4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
8449
CHEBI:16738
2,4-dichlorophenol
OC1=CC=C(Cl)C=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 41°C to 44°C
Color White to Beige
Boiling Point 209°C to 210°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3OH
Quantity 500 g
Beilstein 06, 189
Merck Index 15, 3082
Solubility Information Solubility in water: 4.5g/L in water (20°C). Other solubilities: soluble in carbon tetrachloride,diethyl ether,,benzene,chloroform,ethanol,aromatic solvents,,acetone and sodium hydroxide
Formula Weight 163
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4-Dichlorophenol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichlorophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'