missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dichlorophenol, 99%

Brand:  Alfa Aesar™ A12065.0E

Code : NEW

Additional Details : CAS Number : 120-83-2 Weight : 2.50000kg Transport : UN number : 2928 Chem class : 1 Pack group : II

Quantity 2500g

 View more versions of this product

Product Code. 11419962

  • £117.00 / 2500g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

2,4-Dichlorophenol
C6H4Cl2O
MFCD00002169
742467
120-83-2
163
UN2928
14,3072

Specifications

2,4-Dichlorophenol
209° to 210°C
2500g
99%
Phenol-like
MFCD00002169
742467
114°C (237°F)
40° to 44°C
120-83-2
C6H4Cl2O
163
UN2928
14,3072
Videos
SDS