missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (2,4-Dichlorobenzyl)triphenylphosphonium chloride, 98%

Brand:  Alfa Aesar™ H61405.14

Code : NEW

Additional Details : CAS Number : 2492-23-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15406496

  • £46.85 / 25g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

(2,4-Dichlorobenzyl)triphenylphosphonium chloride
2492-23-1
457.76
MFCD00031641

Specifications

(2,4-Dichlorobenzyl)triphenylphosphonium chloride
249°C
2492-23-1
98%
MFCD00031641
Off-white
25g
Solid
457.76
Videos
SDS