missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl isocyanate, 98%, ACROS Organics™

Brand:  Acros Organics 440350100

Code : 29

Additional Details : CAS Number : 19654-32-1 Weight : 0.06000kg


Quantity 10g

 View more versions of this product

Product Code. 11985691

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2, 4-Dichlorobenzyl isocyanate
97.5
≥97.5% (GC)
C8H5Cl2NO
19654-32-1
100.0
98%
202.04

Specifications

2, 4-Dichlorobenzyl isocyanate
10g
97.5
1.388g/mL
Crystalline Powder
98%
202.04
White to yellow to light brown
19654-32-1
100.0
Authentic
≥97.5% (GC)
C8H5Cl2NO
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4-Dichlorobenzyl isocyanate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS