missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog Number p-7021131
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1777-82-8
C7H6Cl2O
177.024
MFCD00004606
DBHODFSFBXJZNY-UHFFFAOYSA-N
2,4-dichlorobenzyl alcohol, 2,4-dichlorophenyl methanol, dybenal, 2,4-dichlorobenzenemethanol, rapidosept, myacide sp, benzenemethanol, 2,4-dichloro, benzyl alcohol, 2,4-dichloro, unii-1nkx3648j9, 2,4-dichlorobenzylalcohol
15684
CHEBI:48220
(2,4-dichlorophenyl)methanol
C1=CC(=C(C=C1Cl)Cl)CO
This item is not returnable. View return policy
1777-82-8
C7H6Cl2O
177.024
MFCD00004606
DBHODFSFBXJZNY-UHFFFAOYSA-N
2,4-dichlorobenzyl alcohol, 2,4-dichlorophenyl methanol, dybenal, 2,4-dichlorobenzenemethanol, rapidosept, myacide sp, benzenemethanol, 2,4-dichloro, benzyl alcohol, 2,4-dichloro, unii-1nkx3648j9, 2,4-dichlorobenzylalcohol
15684
CHEBI:48220
(2,4-dichlorophenyl)methanol
C1=CC(=C(C=C1Cl)Cl)CO

2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

Solubility
Soluble in water, methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 57°C to 61°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1448652
Merck Index 14,3060
Solubility Information Soluble in water,methanol and chloroform.
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,4-Dichlorobenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'