missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzotrifluoride, 99%, ACROS Organics™

Brand:  Acros Organics 166550250

Code : 29

Additional Details : CAS Number : 320-60-5 Weight : 0.07500kg Transport : UN number : 3265 Chem class : 8 Pack group : II


Quantity 25g
Packaging Glass Bottle

 View more versions of this product

Product Code. 10313672

  • £8.60 / 25g


EU Stock 5
Add to basket


Specifications

Chemical Identifiers

2, 4-Dichlorobenzotrifluoride
320-60-5
100.0
99%
Cl2C6H3CF3
MFCD00000582
99%
98.5
≥98.5%
C7H3Cl2F3
215
2, 4-DCBTF, 2, 4-Dichloro-α, α, 2,4-Dichloro-α,α,α-trifluorotoluene

Specifications

2, 4-Dichlorobenzotrifluoride
25g
320-60-5
100.0
Authentic
1.4805 to 1.4825 (20°C, 589nm)
≥98.5%
C7H3Cl2F3
215
2, 4-DCBTF, 2, 4-Dichloro-α, α, 2,4-Dichloro-α,α,α-trifluorotoluene
Clear colorless
99%
98.5
1.4840g/mL
Glass Bottle
Liquid
99%
Cl2C6H3CF3
MFCD00000582
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4-Dichlorobenzotrifluoride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS