missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate, 97%

Brand:  Alfa Aesar™ H55721.03

Code : NEW

Additional Details : CAS Number : 123333-91-5 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15442246

  • £29.36 / 1g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate
461.29 (Anhydrous)
UN3261
123333-91-5
MFCD08457580

Specifications

2,4-Dichlorobenzenediazonium 1,5-naphthalenedisulfonate hydrate
Yellow to pale brown
1g
Powder
461.29 (Anhydrous)
UN3261
Hygroscopic
195°C (dec.)
123333-91-5
97%
MFCD08457580
Videos
SDS