missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzamidoxime, 97%, Thermo Scientific™

Catalog Number p-7033543
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
22179-80-2
C7H6Cl2N2O
205.038
MFCD00178762
MANQHFUAAAGDIX-UHFFFAOYSA-N
2,4-dichlorobenzamidoxime, 2,4-dichloro-n'-hydroxybenzimidamide, 2,4-dichloro-n'-hydroxybenzene-1-carboximidamide, 2,4-dichloro-n-hydroxybenzenecarboximidamide, 2,4-dichlorophenyl hydroxyimino methylamine, z-2,4-dichloro-n'-hydroxybenzimidamide, benzenecarboximidamide,2,4-dichloro-n-hydroxy, hcmhpngpbebdfyyui``xhacp, 2,4-dichloro-n-hydroxybenzimidamide, 2,4-bis chloranyl-n'-oxidanyl-benzenecarboximidamide
9581031
2,4-dichloro-N'-hydroxybenzenecarboximidamide
C1=CC(=C(C=C1Cl)Cl)C(=NO)N
This item is not returnable. View return policy
22179-80-2
C7H6Cl2N2O
205.038
MFCD00178762
MANQHFUAAAGDIX-UHFFFAOYSA-N
2,4-dichlorobenzamidoxime, 2,4-dichloro-n'-hydroxybenzimidamide, 2,4-dichloro-n'-hydroxybenzene-1-carboximidamide, 2,4-dichloro-n-hydroxybenzenecarboximidamide, 2,4-dichlorophenyl hydroxyimino methylamine, z-2,4-dichloro-n'-hydroxybenzimidamide, benzenecarboximidamide,2,4-dichloro-n-hydroxy, hcmhpngpbebdfyyui``xhacp, 2,4-dichloro-n-hydroxybenzimidamide, 2,4-bis chloranyl-n'-oxidanyl-benzenecarboximidamide
9581031
2,4-dichloro-N'-hydroxybenzenecarboximidamide
C1=CC(=C(C=C1Cl)Cl)C(=NO)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 2,4-Dichlorobenzamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'