missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-α-methylbenzyl alcohol, 99+%, Thermo Scientific™

Catalog Number 10503751
Click to view available options
10g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 1475-13-4
CAS Min % 99.0
CAS Max % 100.0
Density 1.2900g/mL
Boiling Point 125.0°C to 126.0°C (7.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C8H8Cl2O
Linear Formula Cl2C6H3CH(CH3)OH
Refractive Index 1.5590 to 1.5610
MDL Number MFCD00004511
Quantity 10g
Beilstein 06, II, 446
Specific Gravity 1.29
Synonym 1-2,4-dichlorophenyl ethanol, 1-2,4-dichlorophenyl ethan-1-ol, 2,4-dichloro-alpha-methylbenzyl alcohol, 1-2,4-dichloro-phenyl ethanol, 1-2,4-dichlorophenyl ethyl alcohol, 2,4-dichloro-alpha-methylbenzylalcohol, 2,4-dichloro-1-1-hydroxyethyl benzene, 2,4-dichloro-alpha-methylbenzenemethanol, 2,4-dichloro-.alpha.-methylbenzyl alcohol, 2,4-dichloro-a-methylbenzyl alcohol
InChI Key KWZDYNBHZMQRLS-UHFFFAOYSA-N
SMILES CC(C1=C(C=C(C=C1)Cl)Cl)O
IUPAC Name 1-(2,4-dichlorophenyl)ethanol
Molecular Weight (g/mol) 191.06
PubChem CID 92889
Formula Weight 191.06
Percent Purity 99+%
Chemical Name or Material 2,4-Dichloro-α-methylbenzyl alcohol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichloro-a-methylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'