missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-α-methylbenzyl alcohol, 99+%, ACROS Organics™

Brand:  Acros Organics 188970500

Code : 29

Additional Details : CAS Number : 1475-13-4 Weight : 0.10000kg


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10377890

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2, 4-Dichloro-α-methylbenzyl alcohol
1475-13-4
100.0
99+%
Cl2C6H3CH(CH3)OH
MFCD00004511
99+%
99.0
≥99.0%
C8H8Cl2O
191.06

Specifications

2, 4-Dichloro-α-methylbenzyl alcohol
Clear colorless
99+%
99.0
Authentic
≥99.0%
C8H8Cl2O
191.06
Glass bottle
06, II, 446
1.29g/mL
50g
1475-13-4
100.0
Liquid
99+%
Cl2C6H3CH(CH3)OH
MFCD00004511
1.5590 to 1.5610 (20°C, 589nm)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4-Dichloro-a-methylbenzyl alcohol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS