missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 250°C to 252°C |
Assay Percent Range | 94% min. (HPLC) |
Quantity | 250 mg |
Formula Weight | 188.02 |
Percent Purity | 95% |
Chemical Name or Material | 2, 4-Dichloro-7H-pyrrolo[2, 3-d]pyrimidine |
Safety and Handling
Product Identifier
- 2,4-Dichloro-7H-pyrrolo¢2,3-d!pyrimidine, 95%
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'