missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dichloro-6-methylphenol, 98%

Brand:  Alfa Aesar™ H64312.06

Code : NEW

Additional Details : CAS Number : 1570-65-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15471459

  • £164.30 / 5g


Estimated Shipment date
from Supplier 19-03-2018
Add to basket

Specifications

Chemical Identifiers

2,4-Dichloro-6-methylphenol
177.02
UN3077
1570-65-6
MFCD00019988
1864865

Specifications

2,4-Dichloro-6-methylphenol
Brown
5g
Solid
177.02
UN3077
225°C (lit.)
52° to 56°C
1570-65-6
98%
MFCD00019988
1864865
Videos
SDS