missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-nitropyrimidine, 97%, ACROS Organics™

Brand:  Acros Organics 435290010

Code : 29

Additional Details : CAS Number : 49845-33-2 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10658360

  • £15.60 / 1g


Estimated Shipment
23-03-2018
Add to basket


Specifications

Chemical Identifiers

2, 4-Dichloro-5-nitropyrimidine
96.0
≥96.0%
C4HCl2N3O2
MFCD00127867
49845-33-2
100.0
97%
193.98

Specifications

2, 4-Dichloro-5-nitropyrimidine
28° to 32°C
49845-33-2
96.0
Glass bottle
≥96.0%
C4HCl2N3O2
MFCD00127867
Clear yellow
1g
Authentic
100.0
Liquid or Low Melting Solid
97%
193.98
Videos
SDS