missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-nitropyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number 10535100
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
49845-33-2
C4HCl2N3O2
193.97
MFCD00127867
INUSQTPGSHFGHM-UHFFFAOYSA-N
2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine
521266
2,4-dichloro-5-nitropyrimidine
[O-][N+](=O)C1=CN=C(Cl)N=C1Cl
This item is not returnable. View return policy
49845-33-2
C4HCl2N3O2
193.97
MFCD00127867
INUSQTPGSHFGHM-UHFFFAOYSA-N
2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine
521266
2,4-dichloro-5-nitropyrimidine
[O-][N+](=O)C1=CN=C(Cl)N=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 29°C to 30°C
Color Yellow
Boiling Point 135°C (17.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 193.98
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2, 4-Dichloro-5-nitropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichloro-5-nitropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'