missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number 15411378
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1780-31-0
C5H4Cl2N2
163.001
MFCD00023197
DQXNTSXKIUZJJS-UHFFFAOYSA-N
pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p
74508
2,4-dichloro-5-methylpyrimidine
CC1=CN=C(N=C1Cl)Cl
This item is not returnable. View return policy
1780-31-0
C5H4Cl2N2
163.001
MFCD00023197
DQXNTSXKIUZJJS-UHFFFAOYSA-N
pyrimidine, 2,4-dichloro-5-methyl, 5-methyl-2,4-dichloropyrimidine, 2,4-dichloro-5-methyl-pyrimidine, 2,4-dichloro-5-methyl pyrimidine, 2, 4-dichloro-5-methylpyrimidine, 2,4-dichlor-5-methylpyrimidin, pubchem5288, zlchem 1254, acmc-1bsov, ksc178k6p
74508
2,4-dichloro-5-methylpyrimidine
CC1=CN=C(N=C1Cl)Cl

2,4-Dichloro-5-methylpyrimidine, is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine. It can react with piperidineto produce 2-chloro-5-methyl-4-piperidin-1-yl-pyrimidine. This reaction will need solvent dioxane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloro-5-methylpyrimidine, is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine. It can react with piperidineto produce 2-chloro-5-methyl-4-piperidin-1-yl-pyrimidine. This reaction will need solvent dioxane.

Solubility
Soluble in chloroform, ether, ethyl acetate and toluene

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 26°C to 28°C
Density 1.39
Boiling Point 108°C to 109°C (11 mmHg)
Flash Point 113°C (235°F)
Quantity 250 mg
UN Number UN3261
Solubility Information Soluble in chloroform,ether,ethyl acetate and toluene
Formula Weight 163.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-methylpyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'