missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methylpyridine, 96%, Thermo Scientific™

Catalog Number p-7053887
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
56961-78-5
C6H5Cl2N
162.01
MFCD08704813
DUPJNQNLAKJWKU-UHFFFAOYSA-N
2,4-dichloro-5-picoline, pyridine, 2,4-dichloro-5-methyl, 2,4-dichloro-5-methyl-pyridine, pubchem5464, acmc-209lv3, ksc495i9n, 2,4-dichloro-5-methyl pyridine, 2,4-bis chloranyl-5-methyl-pyridine
12643718
2,4-dichloro-5-methylpyridine
CC1=CN=C(Cl)C=C1Cl
This item is not returnable. View return policy
56961-78-5
C6H5Cl2N
162.01
MFCD08704813
DUPJNQNLAKJWKU-UHFFFAOYSA-N
2,4-dichloro-5-picoline, pyridine, 2,4-dichloro-5-methyl, 2,4-dichloro-5-methyl-pyridine, pubchem5464, acmc-209lv3, ksc495i9n, 2,4-dichloro-5-methyl pyridine, 2,4-bis chloranyl-5-methyl-pyridine
12643718
2,4-dichloro-5-methylpyridine
CC1=CN=C(Cl)C=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4-Dichloro-5-methylpyridine is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 162.02
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2,4-Dichloro-5-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'