missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dichloro-5-(iodomethyl)pyrimidine, 98%

Brand:  Alfa Aesar™ H64805.MD

Code : NEW

Additional Details : CAS Number : 7627-44-3 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15482169

  • £64.77 / 250mg


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

2,4-Dichloro-5-(iodomethyl)pyrimidine
288.9
512189
7627-44-3
MFCD12827832

Specifications

2,4-Dichloro-5-(iodomethyl)pyrimidine
52° to 53°C
7627-44-3
98%
MFCD12827832
Light Sensitive
250mg
Solid
288.9
512189
Videos
SDS