missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-fluorobenzamide, 97+%, Thermo Scientific™

Catalog Number p-7027560
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy
135748-33-3
C7H4Cl2FNO
208.01
MFCD01631478
SKSVSZRFINHARI-UHFFFAOYSA-N
acmc-20a3pc, 2,4-dichloro-5-fluoro-benzamide, benzamide, 2,4-dichloro-5-fluoro, 2,4-dichloro-5-fluorobenzamide
2774011
2,4-dichloro-5-fluorobenzamide
NC(=O)C1=CC(F)=C(Cl)C=C1Cl
This item is not returnable. View return policy
135748-33-3
C7H4Cl2FNO
208.01
MFCD01631478
SKSVSZRFINHARI-UHFFFAOYSA-N
acmc-20a3pc, 2,4-dichloro-5-fluoro-benzamide, benzamide, 2,4-dichloro-5-fluoro, 2,4-dichloro-5-fluorobenzamide
2774011
2,4-dichloro-5-fluorobenzamide
NC(=O)C1=CC(F)=C(Cl)C=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 158°C to 160°C
Quantity 1 g
Formula Weight 208.02
Percent Purity ≥97%
Chemical Name or Material 2,4-Dichloro-5-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'