missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dichloro-3-fluorobenzonitrile, 98%

Brand:  Alfa Aesar™ H61705.03

Code : NEW

Additional Details : CAS Number : 161612-68-6 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15498268

  • £71.76 / 1g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

2,4-Dichloro-3-fluorobenzonitrile
190
UN3439
161612-68-6
MFCD06658268

Specifications

2,4-Dichloro-3-fluorobenzonitrile
1g
Crystalline
190
UN3439
Off-white to pale yellow
161612-68-6
98%
MFCD06658268
Videos
SDS