missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dichloro-1-(4-nitrophenoxy)benzene, 98%

£31.27 - £122.60

Chemical Identifiers

CAS 1836-75-5
Molecular Formula C12H7Cl2NO3
Molecular Weight (g/mol) 284.092
MDL Number MFCD00128026
InChI Key XITQUSLLOSKDTB-UHFFFAOYSA-N
Synonym nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
PubChem CID 15787
IUPAC Name 2,4-dichloro-1-(4-nitrophenoxy)benzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11448857 SDS View Documents Alfa Aesar™
A19977.06
5g £31.27
5g View alternative products
Save Up to 
»
In stock, ready to ship: 10 Please to purchase this item. Need a web account? Register with us today!
11458857 SDS View Documents Alfa Aesar™
A19977.14
25g £122.60
25g View alternative products
Save Up to 
»
In stock, ready to ship: 5 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1836-75-5
284.092
XITQUSLLOSKDTB-UHFFFAOYSA-N
15787
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl
C12H7Cl2NO3
MFCD00128026
nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
2,4-dichloro-1-(4-nitrophenoxy)benzene

Specifications

2,4-Dichloro-1-(4-nitrophenoxy)benzene
5g
C12H7Cl2NO3
UN3077
14,6598
XITQUSLLOSKDTB-UHFFFAOYSA-N
2,4-dichloro-1-(4-nitrophenoxy)benzene
15787
98%
68°C to 71°C
1836-75-5
MFCD00128026
1887356
nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl
284.092
284.1