missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-1-(4-nitrophenoxy)benzene, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Density | 1.3000g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | Cl2C6H3OC6H4NO2 |
Quantity | 25 g |
Formula Weight | 284.1 |
Percent Purity | 97% |
Chemical Name or Material | 2, 4-Dichloro-1-(4-nitrophenoxy)benzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'