missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dibromotoluene, 98%

Brand:  Alfa Aesar™ H63869.03

Code : NEW

Additional Details : CAS Number : 31543-75-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15460809

  • £9.80 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

2,4-Dibromotoluene
249.93
2078741
31543-75-6
MFCD00052985
2,4-Dibromo-1-methylbenzene

Specifications

2,4-Dibromotoluene
1.601
98%
MFCD00052985
1.85g/mL
−10°C (dec.)
1g
31543-75-6
Liquid
249.93
2078741
2,4-Dibromo-1-methylbenzene
243°C (dec.)
Videos
SDS