missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromothiophene, 90+%, Thermo Scientific Chemicals

Catalog Number p-7046244
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3140-92-9
C4H2Br2S
241.928
MFCD00043889
WAQFYSJKIRRXLP-UHFFFAOYSA-N
thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64
2724560
2,4-dibromothiophene
C1=C(SC=C1Br)Br
This item is not returnable. View return policy
3140-92-9
C4H2Br2S
241.928
MFCD00043889
WAQFYSJKIRRXLP-UHFFFAOYSA-N
thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64
2724560
2,4-dibromothiophene
C1=C(SC=C1Br)Br

2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Density 2.16
Boiling Point 215°C
Flash Point >110°C (230°F)
Refractive Index 1.632
Quantity 1 g
UN Number UN2810
Beilstein 107651
Solubility Information It is insoluble in water.
Formula Weight 241.94
Percent Purity ≥90%
Chemical Name or Material 2,4-Dibromothiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'