missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenol 99%, ACROS Organics™

Brand:  Acros Organics 406511000

Code : 29

Additional Details : CAS Number : 615-58-7 Weight : 0.16000kg Transport : UN number : 2811 Chem class : 1 Pack group : II

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10792582

  • £77.50 / 100g


Estimated Shipment
16-05-2018
Add to basket

Specifications

Chemical Identifiers

2, 4-Dibromophenol
615-58-7
100.0
99%
Br2C6H3OH
MFCD00002149
99%
98.5
≥98.5%
C6H4Br2O
251.91

Specifications

2, 4-Dibromophenol
36° to 39°C
99%
Authentic
100.0
Crystalline Mass, Powder, Crystals and/or Chunks
99%
Br2C6H3OH
MFCD00002149
White to slightly yellow or beige
100g
615-58-7
98.5
Glass bottle
≥98.5%
C6H4Br2O
251.91
Videos
SDS