missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dibromobenzyl alcohol, 98%

Brand:  Alfa Aesar™ H64741.03

Code : NEW

Additional Details : CAS Number : 666747-06-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412059

  • £66.20 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Description

Description

Applications
It is an halogenated compound for proteomics research. Also used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2,4-Dibromobenzyl alcohol
265.98
3241294
666747-06-4
MFCD09996906

Specifications

2,4-Dibromobenzyl alcohol
Solid
1g
MFCD09996906
666747-06-4
98%
265.98
3241294
Videos
SDS