missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dibromobenzyl alcohol, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an halogenated compound for proteomics research. Also used as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It is an halogenated compound for proteomics research. Also used as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Quantity | 5 g |
Beilstein | 3241294 |
Synonym | 2,4-dibromophenyl methanol, 2,4-dibromobenzyl alcohol, benzenemethanol, 2,4-dibromo, 2,4-dibromobenzenemethanol, benzenemethanol,2,4-dibromo, pubchem9781, 2,4-dibromobenzylalcohol, acmc-1bfu5, 2,4-bis bromanyl phenyl methanol |
Solubility Information | Slightly soluble in water. |
Formula Weight | 265.98 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2,4-Dibromobenzyl alcohol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'