missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromoanisole, 98%, Thermo Scientific™

Catalog Number p-102712
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
21702-84-1
C7H6Br2O
265.92
MFCD00000079
XGXUGXPKRBQINS-UHFFFAOYSA-N
2,4-dibromoanisole, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, pubchem3847, acmc-209fnf, maybridge1_003667, chemwish ic06728, 2,4-dibromo-1-methoxy-benzene, xgxugxpkrbqins-uhfffaoysa, #
27011
2,4-dibromo-1-methoxybenzene
COC1=C(C=C(C=C1)Br)Br
This item is not returnable. View return policy
21702-84-1
C7H6Br2O
265.92
MFCD00000079
XGXUGXPKRBQINS-UHFFFAOYSA-N
2,4-dibromoanisole, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, pubchem3847, acmc-209fnf, maybridge1_003667, chemwish ic06728, 2,4-dibromo-1-methoxy-benzene, xgxugxpkrbqins-uhfffaoysa, #
27011
2,4-dibromo-1-methoxybenzene
COC1=C(C=C(C=C1)Br)Br
CAS Min % 97.5
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color Beige
Boiling Point 101.0°C to 106.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br2(C6H3)OCH3
Quantity 5 g
Formula Weight 265.92
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dibromoanisole, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'