missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromoanisole, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023119
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
21702-84-1
C7H6Br2O
265.932
MFCD00000079
XGXUGXPKRBQINS-UHFFFAOYSA-N
27011
2,4-dibromo-1-methoxybenzene
COC1=C(C=C(C=C1)Br)Br
This item is not returnable. View return policy
21702-84-1
C7H6Br2O
265.932
MFCD00000079
XGXUGXPKRBQINS-UHFFFAOYSA-N
27011
2,4-dibromo-1-methoxybenzene
COC1=C(C=C(C=C1)Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 60°C to 64°C
Boiling Point 82°C to 88°C (0.3 mmHg)
Quantity 25 g
Beilstein 2045290
Synonym 2,4-dibromoanisole, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, pubchem3847, acmc-209fnf, maybridge1_003667, chemwish ic06728, 2,4-dibromo-1-methoxy-benzene, xgxugxpkrbqins-uhfffaoysa, #
Formula Weight 265.93
Percent Purity ≥98%
Chemical Name or Material 2,4-Dibromoanisole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'