missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2',4'-Dibromoacetophenone, 98%

Brand:  Alfa Aesar™ H61181.06

Code : NEW

Additional Details : CAS Number : 33243-33-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407688

  • £376.30 / 5g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2',4'-Dibromoacetophenone
33243-33-3
277.94
MFCD00152057

Specifications

2',4'-Dibromoacetophenone
Solid
5g
MFCD00152057
33243-33-3
98%
277.94
Videos
SDS