missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dibromo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65293.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409546

  • £59.78 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

2,4-Dibromo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
409.12
MFCD28098779

Specifications

2,4-Dibromo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
Solid
409.12
1g
95%
MFCD28098779
Videos
SDS