missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-7036004
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
175401-95-3
C15H9N3O2
263.256
MFCD20265213
HRUUSKALOILBKF-UHFFFAOYSA-N
2-4-cyanophenyl benzimidazole-6-carboxylic acid, 2-4-cyanophenyl benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-3h-1,3-benzodiazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-6-carboxylic acid, 1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl, 2-4-cyanophenyl-1h-benzoimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzo d imidazole-6-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-1,3-benzodiazole-5-carboxylic acid
23509758
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O
This item is not returnable. View return policy
175401-95-3
C15H9N3O2
263.256
MFCD20265213
HRUUSKALOILBKF-UHFFFAOYSA-N
2-4-cyanophenyl benzimidazole-6-carboxylic acid, 2-4-cyanophenyl benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-3h-1,3-benzodiazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-6-carboxylic acid, 1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl, 2-4-cyanophenyl-1h-benzoimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzo d imidazole-6-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-1,3-benzodiazole-5-carboxylic acid
23509758
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 263.26
Percent Purity 97%
Chemical Name or Material 2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'