missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Chlorophenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62341.03

Code : NEW

Additional Details : CAS Number : 34491-29-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477306

  • £77.30 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Chlorophenyl)thiazolidine-4-carboxylic acid
34491-29-7
243.71
MFCD00278428

Specifications

2-(4-Chlorophenyl)thiazolidine-4-carboxylic acid
1g
Solid
243.71
White
34491-29-7
97%
MFCD00278428
Videos
SDS