missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Chlorophenyl)pyrrolidine, 95%

Brand:  Alfa Aesar™ H61851.03

Code : NEW

Additional Details : CAS Number : 38944-14-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448438

  • £159.20 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Chlorophenyl)pyrrolidine
38944-14-8
181.66
MFCD01631838

Specifications

2-(4-Chlorophenyl)pyrrolidine
38944-14-8
95%
MFCD01631838
1g
Solid
181.66
Videos
SDS