missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)ethylamine 98%, ACROS Organics™

Brand:  Acros Organics 109880050

Code : X2

Additional Details : CAS Number : 156-41-2 Weight : 0.05556kg Transport : UN number : 2735 Chem class : 1b Pack group : III


Quantity 5mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10403845

  • £15.60 / 5mL


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

2-(4-Chlorophenyl)ethylamine
156-41-2
100.0
98%
ClC6H4CH2CH2NH2
MFCD00008191
98%
97.5
98%
C8H10ClN
155.63
4-Chlorophenethylamine

Specifications

2-(4-Chlorophenyl)ethylamine
106°C
Clear colorless to yellow
98%
97.5
Authentic
Glass bottle
Liquid
98%
ClC6H4CH2CH2NH2
MFCD00008191
4-Chlorophenethylamine
1.1120g/mL
242.0° to 245.0°C
5mL
156-41-2
100.0
1.5474 to 1.5494
0.5% max. (K.F.)
98%
C8H10ClN
155.63
12, IV, 2489
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(4-Chlorophenyl)ethylamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

Documentation

Technical Documents