missing translation for 'onlineSavingsMsg'
Learn More

(2-(4-Chlorophenyl)-1,3-thiazol-4-yl)methanamine hydrochloride, 97%, Tech., Thermo Scientific™

Catalog Number p-3075147
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
690632-35-0
C10H10Cl2N2S
261.16
MFCD05865127
PNBZPHKDYJHRSP-UHFFFAOYSA-N
2-4-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride, 2-4-chlorophenyl thiazol-4-yl methanamine hydrochloride, 2-4-chlorophenyl-1,3-thiazol-4-yl methanaminehydrochloride, bestipharma 584-709, 2-4-chloro-phenyl-thiazol-4-ylmethylamine hcl, 2-4-chlorophenyl-4-thiazolyl methanamine hydrochloride, 2-4-chlorophenyl-1,3-thiazol-4-yl methylamine, chloride, 2-4-chloro-phenyl-thiazol-4-ylmethylamine hydrochloride, 1-2-4-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride
2794737
Cl.NCC1=CSC(=N1)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
690632-35-0
C10H10Cl2N2S
261.16
MFCD05865127
PNBZPHKDYJHRSP-UHFFFAOYSA-N
2-4-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride, 2-4-chlorophenyl thiazol-4-yl methanamine hydrochloride, 2-4-chlorophenyl-1,3-thiazol-4-yl methanaminehydrochloride, bestipharma 584-709, 2-4-chloro-phenyl-thiazol-4-ylmethylamine hcl, 2-4-chlorophenyl-4-thiazolyl methanamine hydrochloride, 2-4-chlorophenyl-1,3-thiazol-4-yl methylamine, chloride, 2-4-chloro-phenyl-thiazol-4-ylmethylamine hydrochloride, 1-2-4-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride
2794737
Cl.NCC1=CSC(=N1)C1=CC=C(Cl)C=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name [2-(4-chlorophenyl)-1,3-thiazol-4-yl]methanamine;hydrochloride
Formula Weight 261.18
Percent Purity 97%
Grade Technical
Chemical Name or Material [2-(4-Chlorophenyl)-1, 3-thiazol-4-yl]methanamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'