missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, ACROS Organics™

Brand:  Acros Organics 189090500

Code : 29

Additional Details : CAS Number : 882-09-7 Weight : 0.10000kg

Quantity 50g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10205480

  • £24.00 / 50g


EU Stock 4
Add to basket

Specifications

Chemical Identifiers

2-(4-Chlorophenoxy)-2-methylpropionic acid
882-09-7
100.0
99%
ClC6H4OC(CH3)2CO2H
MFCD00004192
99%
98.5
99%
C10H11ClO3
214.65
Clofibric acid

Specifications

2-(4-Chlorophenoxy)-2-methylpropionic acid
120.0° to 123.0°C
99%
98.5
Authentic
Plastic bottle
99%
C10H11ClO3
214.65
15, 2371
Pale yellow to brownish
50g
882-09-7
100.0
1% max.
Crystals or Crystalline Powder
99%
ClC6H4OC(CH3)2CO2H
MFCD00004192
Clofibric acid
Videos
SDS