missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-90388
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
882-09-7
C10H11ClO3
214.65
MFCD00004192
TXCGAZHTZHNUAI-UHFFFAOYSA-N
clofibric acid, 2-4-chlorophenoxy-2-methylpropanoic acid, chlorofibrinic acid, clofibrinic acid, chlorfibrinic acid, clofibrin, chlorophibrinic acid, pcib, 2-4-chlorophenoxy-2-methylpropionic acid, clofibrate free acid
2797
CHEBI:34648
2-(4-chlorophenoxy)-2-methylpropanoic acid
CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl
This item is not returnable. View return policy
882-09-7
C10H11ClO3
214.65
MFCD00004192
TXCGAZHTZHNUAI-UHFFFAOYSA-N
clofibric acid, 2-4-chlorophenoxy-2-methylpropanoic acid, chlorofibrinic acid, clofibrinic acid, chlorfibrinic acid, clofibrin, chlorophibrinic acid, pcib, 2-4-chlorophenoxy-2-methylpropionic acid, clofibrate free acid
2797
CHEBI:34648
2-(4-chlorophenoxy)-2-methylpropanoic acid
CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 120.0°C to 123.0°C
Color Brown to Yellow
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula ClC6H4OC(CH3)2CO2H
Quantity 250 g
Merck Index 15, 2371
Formula Weight 214.65
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'